Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (OX-38) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280648
Description
CD4 Monoclonal specifically detects CD4 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
CD4 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:100-1:200, Immunohistochemistry : 1:10-1:500, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Frozen : 1:10-1:500, CyTOF-ready | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reacts with Rat. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF | |
OX-38 | |
Unconjugated | |
PBS | |
CD4 | |
MLR generated rat T cells | |
0.25 mg | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
920 | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction