Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (V46P5B10*D5), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250140AF405
Description
CD4 Monoclonal specifically detects CD4 in Fish samples. It is validated for ELISA, Product Image.Specifications
CD4 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD4 | |
Ovalbumin-conjugated synthetic peptide | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
920 | |
Fish | |
Purified |
ELISA | |
V46P5B10*D5 | |
ELISA | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Specific for three-spined stickleback CD4. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction