Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (W3/25), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280647AF594
Description
CD4 Monoclonal antibody specifically detects CD4 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Cell Surface)Specifications
| CD4 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| W3/25 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface) | |
| CD_antigen: CD4, CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Rat Thymocyte Membrane Glycoproteins | |
| 0.25 mL | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
| 920 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction