Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (YNB46.1.8 (Campath-9H)), DyLight 680, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP275900FR
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD4 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
CD4 | |
Rat T-cell line (NB2) transfected with the human CD4 | |
0.1 mL | |
Primary | |
Recognizes the 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes. | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
YNB46.1.8 (Campath-9H) | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Rat | |
Protein A purified | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
920.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction