Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD4/CD25 Antibody Cocktail, FITC, PE, eBioscience™

Catalog No. 5015421
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-154-21 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-154-21 Supplier Invitrogen™ Supplier No. 22842571
Add to Cart
Add to Cart

Antibody Cocktail

Description: This product consists of two fluorochrome-conjugated monoclonal antibodies to identify CD4+CD25+ human peripheral blood cells. The RPA-T4 monoclonal antibody (mouse IgG1, kappa) reacts with human CD4, a 59 kDa cell surface receptor that is expressed by a majority of thymocytes, a subpopulation of mature T cells (T helper cells) and at low levels on monocytes. The BC96 monoclonal antibody (mouse IgG1, kappa) reacts with human CD25, the 55 kDa interleukin-2 receptor alpha chain (IL-2Ralpha). This cocktail of RPA-T4 and BC96 can be used for the flow cytometric detection of human regulatory T cells, which also express Foxp3. Applications Reported: This product has been reported for use in flow cytometric analysis. Applications Tested: This cocktail has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 20 μL per test (containing 1 μg of Anti-Human CD4 (RPA-T4) FITC and 0.125 μg of Anti-Human CD25 (BC96) PE). A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

The CD4 antigen is involved in the recognition of the type II MHC antigen. It is also a receptor for HIV. It is present on most T helper cells and normal thymocytes. The cytoplasmic portion of CD4 is associated with p56lck tyrosine kinase. CD4 expression is commonly found in human lymph nodes and tonsils. CD44 is a type 1 transmembrane glycoprotein also known as Phagocytic Glycoprotein-1 (pgp-1) and HCAM. CD44 is the receptor for hyaluronate and exists as a large number of different isoforms due to alternative RNA splicing. The major isoform expressed on lymphocytes, myeloid cells, and erythrocytes is a glycosylated type 1 transmembrane protein. Other isoforms contain glycosaminoglycans and are expressed on hematopoietic and non-hematopoietic cells. CD44 is involved in adhesion of leukocytes to endothelial cells, stromal cells, and the extracellular matrix. CD25: IL2 receptor alpha (IL2RA) and beta (IL2RB) chains, together with the common gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric alpha chains (IL2RA) result in low-affinity receptor, while homodimeric beta (IL2RB) chains produce a medium-affinity receptor. Normally an integral-membrane protein, soluble IL2RA has been isolated and determined to result from extracellular proteolyisis. Alternately-spliced IL2RA mRNAs have been isolated, but the significance of each is presently unknown. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4/CD25
Applications Flow Cytometry
Classification Cocktail
Clone RPA-T4, BC96
Concentration 20 μL/Test
Conjugate FITC/PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CD4
Gene Accession No. P01589, P01730
Gene Alias CD25; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 receptor; CD4mut; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; IL-2 receptor alpha subunit; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; Il2r; IL-2R alpha; IL-2R alpha chain; IL2R subunit alpha; IL-2R subunit alpha; Il2ra; IL-2-RA; IL2-RA; Il2ra antibody; IL2RAC; IMD41; interleukin 2 receptor subunit alpha; interleukin 2 receptor, alpha; interleukin 2 receptor, alpha chain; interleukin-2 receptor alpha chain; Interleukin2 receptor subunit alpha; interleukin-2 receptor subunit alpha; L3T4; Ly-4; Ly-43; p55; p55 chain; sIL 2R; soluble IL 2 receptor; TAC antigen; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); TCGFR; W3/25; W3/25 antigen
Gene Symbols CD4, IL2RA
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3559, 920
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.