Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD4/CD8 Antibody Cocktail, FITC, PE
GREENER_CHOICE

Catalog No. PIMA517009
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517009 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517009 Supplier Invitrogen™ Supplier No. MA517009
Add to Cart
Add to Cart

Antibody Cocktail

MA5-17009 contains a monoclonal mixture of FITC-conjugated CD4 (clone #YTS191.1, Rat IgG2b) and RPE-conjugated CD8 (clone #KT15, Rat IgG2a). This antibody is a dual-color reagent recognizing mouse CD4 and CD8 cell surface antigens. Clone YTS191.1 recognizes murine CD4, a T cell differentiation antigen expressed on thymocytes and helper/inducer T cells in the peripheral blood. Clone KT15 recognizes a non-polymorphic epitope on the murine CD8 alpha chain. Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

CD4, a single chain transmembrane glycoprotein, is found on a T cell subset (helper/inducer) representing 45% of peripheral blood lymphocytes. It is also present on 80% of thymocytes and at a lower level on monocytes. It is involved in recognition of antigen presented along with MHC class II by APCs. It serves as receptor for HIV. The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen acts as a corepressor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The coreceptor functions as either a homodimer composed of two alpha chains, or as a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4/CD8
Applications Flow Cytometry
Classification Cocktail
Clone YTS191.1/KT15
Concentration Conc. Not Determined
Conjugate FITC/PE
Formulation PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Gene CD8A
Gene Accession No. P01731, P06332
Gene Alias BB154331; Cd4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 receptor; CD4mut; CD8; CD8 antigen; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide (p32); CD8 antigen, beta polypeptide 1 (p37); CD8A; CD8a molecule; CD8alpha; CD8b; CD8b molecule; CD8B1; CD8beta; L3T4; LEU2; Leu-2; Leu2 T-lymphocyte antigen; Ly-2; LY3; Ly-35; Ly-4; Ly-B; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at 100); LYT3; MAL; OKT8 T-cell antigen; p32; P37; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T8 T-cell antigen; T-cell antigen Leu2; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-2; T-lymphocyte differentiation antigen T8/Leu-2
Gene Symbols CD4, CD8A
Host Species Rat
Immunogen Clone YTS191.1: mouse thymocytes. Clone KT15: V beta 11 + T-cell clone.
Purification Method Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12504, 12525
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Lyophilized
Isotype IgG2a, IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.