Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4/CD8 Antibody Cocktail, Invitrogen™

Catalog No. PIMA514432
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514432 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514432 Supplier Thermo Scientific Supplier No. MA514432
Add to Cart
Add to Cart

Antibody Cocktail

This CD4/CD8 monoclonal cocktail contains Mouse HRP CD4 (Clone 4B12, IgG1 kappa) and Rabbit AP CD8 (Clone SP16, IgG). It is designed for qualitative immunohistochemistry with normal and neoplastic formalin-fixed, paraffin-embedded tissues using the MultiVision anti-mouse/HRP + anti-rabbit/AP detection system (Catalog No. TL-012-MHRA). This cocktail is provided at an optimal concentration in an easy-to-use format for visualizing CD4 and CD8 lymphocyte subsets in red and blue, respectively. The use of rabbit monoclonal CD8 allows for superior staining, in both sensitivity and specificity, permiting the use of higher titers compared to mouse monoclonal antibodies. These cocktails are specially formulated from MultiVision polymers that provide increased sensitivity, time-savings, and detection simplicity. The MultiVision AP and HRP polymers are innovative, patented technology. The smaller amino acid polymer subunits minimize conflicts in binding the target protein resulting in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency as well as better signal-to-noise ratios. This system is biotin-free, which prevents background staining found with traditional biotin-based methods. The MultiVision polymer detection systems yield high quality double staining in less than 2 hours on formalin-fixed and paraffin-embedded tissue sections.

CD4, a single chain transmembrane glycoprotein, is found on a T cell subset (helper/inducer) representing 45% of peripheral blood lymphocytes. It is also present on 80% of thymocytes and at a lower level on monocytes. It is involved in recognition of antigen presented along with MHC class II by APCs. It serves as receptor for HIV. The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen acts as a corepressor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The coreceptor functions as either a homodimer composed of two alpha chains, or as a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4/CD8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Cocktail
Clone 4B12, SP16, CD8-SP16
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.09% sodium azide
Gene CD8B
Gene Accession No. P01730, P01732, P10966
Gene Alias BB154331; Cd4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 receptor; CD4mut; CD8; CD8 antigen; CD8 antigen, alpha chain; CD8 antigen, alpha polypeptide (p32); CD8 antigen, beta polypeptide 1 (p37); CD8A; CD8a molecule; CD8alpha; CD8b; CD8b molecule; CD8B1; CD8beta; L3T4; LEU2; Leu-2; Leu2 T-lymphocyte antigen; Ly-2; LY3; Ly-35; Ly-4; Ly-B; Lyt2; Lyt-2; Lyt-2.1 lymphocyte differentiation antigen (AA at 100); LYT3; MAL; OKT8 T-cell antigen; p32; P37; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T8 T-cell antigen; T-cell antigen Leu2; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-2; T-lymphocyte differentiation antigen T8/Leu-2
Gene Symbols CD4, CD8A, CD8B
Host Species Mixed, Rabbit
Immunogen CD4-stimulated human leukocytes, CD8-synthetic peptide containing the 13 C-terminal amino acids of the cytoplasmic domain of the a-chain of the CD8 molecule
Purification Method purified
Quantity 7 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 920, 925, 926
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.