Learn More
Invitrogen™ CD4/HLA-DR Antibody Cocktail, FITC, PE

Description
This antibody is a monoclonal mixture of FITC-conjugated CD4 (clone 7E14, isotype IgG1) and PE-conjugated HLADR (clone 423L, isotype IgG2a).
Specifications
Specifications
| Antigen | CD4/HLA-DR |
| Applications | Flow Cytometry |
| Classification | Cocktail |
| Clone | 7E14, 423L, 423L |
| Conjugate | FITC/PE |
| Formulation | PBS with 0.2% carrier protein and 0.08% sodium azide |
| Gene | CD4 |
| Gene Accession No. | P01730 |
| Gene Alias | CD4; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 receptor; CD4mut; CD74; CD74 antigen; CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated); CD74 molecule; CD74 molecule, major histocompatibility complex, class II invariant chain; DHLAG; gamma chain of class II antigens; HLA class II histocompatibility antigen gamma chain; HLADG; HLA-DR antigens-associated invariant chain; HLA-DR-gamma; Ia antigen-associated invariant chain; Ia-associated invariant chain; Ia-GAMMA; II; MHC HLA-DR gamma chain; p33; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4 |
| Gene Symbols | CD4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.