Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Antibody (CE9.1 (Clenoliximab)) - Azide and BSA Free, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP2759100.2MG
Description
CD4 Monoclonal specifically detects CD4 in Human, Chimpanzee samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Surface Plasmon Resonance, Block/Neutralize.Specifications
CD4 | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300 | |
CD4 | |
Domain 1 of human CD4. | |
Primary | |
The antibody binds to human CD4 with a Kd of 1.0 nM and an ED50 of 0.2 ug/ml. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Blocking Assay, Neutralization | |
CE9.1 (Clenoliximab) | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Surface Plasmon Resonance, Block/Neutralize | |
CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
Human | |
0.2 mg | |
920 | |
Human, Primate | |
IgG4 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction