Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Monoclonal Antibody (SK3 (SK-3)), NovaFluor™ Blue 510, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$209.00 - $463.00
Specifications
Antigen | CD4 |
---|---|
Clone | SK3 (SK-3) |
Concentration | 2 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H001T03B01A
![]() |
Invitrogen™
H001T03B01A |
100 Tests |
Each for $463.00
|
|
|||||
H001T02B01A
![]() |
Invitrogen™
H001T02B01A |
25 Tests |
Each for $209.00
|
|
|||||
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow CytometrySpecifications
CD4 | |
2 μL/Test | |
Monoclonal | |
Mouse | |
Human | |
P01730 | |
920 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
SK3 (SK-3) | |
Flow Cytometry | |
NovaFluor Blue 510 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen | |
IgG1 κ | |
Affinity Chromatography | |
CD4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title