Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Human, APC-Cy7, Clone: SK3 (also known as Leu3a), BD

Catalog No. BDB566319 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566319 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566319 Supplier BD Biosciences Supplier No. 566319
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The SK3 monoclonal antibody specifically binds to CD4. The CD4 antigen is a 55 kDa type 1 transmembrane glycoprotein that belongs to the immunoglobulin superfamily. It is expressed on T-helper/inducer lymphocytes and monocytes. CD3+CD4+ T cells comprise approximately 28% to 58% of normal peripheral blood lymphocytes. CD4 is also expressed on 80% to 95% of normal thymocytes. The CD4 antigen is present in low density on the cell surface of monocytes and in the cytoplasm of monocytes and macrophages (CD3-CD4+). CD4 serves as a T cell coreceptor for MHC class II antigen recognition and as a receptor for the human immunodeficiency virus (HIV). The SK3 antibody inhibits HIV binding to CD4+ cells.

APC-Cy™7 is a tandem fluorochrome that combines APC and a cyanine dye (Cy7). Special precautions must be taken with APC-Cy7 conjugates, and cells stained with them, to protect the fluorochrome from long-term exposure to light. Some APC-Cy7 conjugates show changes in their emission spectra with prolonged exposure to paraformaldehyde. Fixed cells should be analyzed within 4 hours of fixation in paraformaldehyde or transferred to a paraformaldehyde-free buffer for overnight storage. Due to nearly identical excitation and emission properties but different spillover characteristics, APC-Cy7 and APC-H7 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Classification Monoclonal
Clone SK3 (also known as Leu3a)
Conjugate APC-Cyanine7
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Gene Alias L3T4, Leu3a, T-cell surface antigen T4/Leu-3, W3/25, CD4 antigen (p55)
Host Species Mouse
Immunogen Human Peripheral Blood T Cells
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.