Learn More
CD4 Mouse anti-Human, Brilliant Violet 605, Clone: M-T477, BD Optibuild™
Shop All BD Biosciences ProductsDescription
Clone M-T477 reacts with the human form of a 56 kDa transmembrane glycoprotein, CD4, present on the T-helper/inducer subset of normal human donor peripheral blood lymphocytes. This clone also cross-reacts with a subset of CD3-positive peripheral blood lymphoyctes, but not monocytes, of both rhesus and cynomolgus macaque monkeys. Cross-reactivity on both lymphocytes and monocytes (weak) of baboon is also observed. The distribution of M-T477 reactivity on lymphocytes is similar for both human and monkey, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers.
Specifications
Specifications
| Antigen | CD4 |
| Classification | Monoclonal |
| Clone | M-T477 |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 605 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.