Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Mouse anti-Human, BUV563, Clone: SK3, BD Horizon™
Mouse Monoclonal Antibody
$249.00 - $472.00
Specifications
Antigen | CD4 |
---|---|
Clone | SK3 (also known as Leu3a) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 563 |
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry.Specifications
CD4 | |
Flow Cytometry | |
Brilliant Ultraviolet 563 | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
SK3 (also known as Leu3a) | |
Monoclonal | |
Mouse | |
Human | |
Primary | |
CD4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title