Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Mouse anti-Human, BV480, Clone: M-T477, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 746671
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry.
Specifications
CD4 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
M-T477 | |
Brilliant Violet 480 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction