Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Human, Clone: 5D9, Abnova™

Catalog No. 89933809 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-809 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-809 Supplier Abnova Corporation Supplier No. MAB12479
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full length recombinant human CD4.

Specifications

Antigen CD4
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 5D9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human CD4.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.3 (1% BSA, 50% glycerol, 0.02% sodium azide)
Gene Alias CD4mut
Gene Symbols CD4
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human CD4.
Purification Method Protein A/G purification
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 920
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.