Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Mouse anti-Human, PE, Clone: OKT4, Abnova™
Mouse monoclonal antibody raised against CD4.
Supplier: Abnova Corporation MAB12207
Specifications
CD4 | |
Monoclonal | |
PE | |
Flow Cytometry The optimal working dilution should be determined by the end user. | |
CD4mut | |
Mouse | |
Affinity Chromatography | |
RUO | |
920 | |
Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
IgG2b κ |
Flow Cytometry | |
OKT4 | |
Mouse monoclonal antibody raised against CD4. | |
In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer). | |
CD4 | |
CD4 | |
100 Reactions | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction