Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse anti-Human, FITC, Clone: RPA-T4, Abnova™

Catalog No. 89933804 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-804 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-804 Supplier Abnova Corporation Supplier No. MAB12208
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against CD4.

Specifications

Antigen CD4
Applications Flow Cytometry
Classification Monoclonal
Clone RPA-T4
Conjugate FITC
Description Mouse monoclonal antibody raised against CD4.
Dilution Flow Cytometry The optimal working dilution should be determined by the end user.
Formulation In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer).
Gene Alias CD4mut
Gene Symbols CD4
Host Species Mouse
Immunogen CD4
Purification Method Affinity Chromatography
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 920
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.