Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Mouse anti-Human , PE, Clone: OKT4, BD Biosciences™
Mouse Monoclonal Antibody
$50.20 - $88.00
Specifications
Antigen | CD4 |
---|---|
Clone | OKT4 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow CytometrySpecifications
CD4 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
Primary |
OKT4 | |
Monoclonal | |
Mouse | |
Human | |
CD4 antigen (p55); T-cell surface antigen T4/Leu-3 | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title