Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Mouse anti-Porcine, APC, Clone: 74-12-4, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 451511
Description
SouthernBiotech anti-porcine primary antibodies are available in a variety of conjugated formats including Alexa Fluor.
Specifications
CD4 | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Fresh dd miniature swine thymocytes | |
RUO | |
Pig | |
IgG2b |
Flow Cytometry | |
74-12-4 | |
APC | |
Mouse | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction