Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Mouse Anti-Rat, Brilliant Violet 510, Clone: OX-38, BD Optibuild™

Catalog No. BDB743089 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743089 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743089 Supplier BD Biosciences Supplier No. 743089
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The OX-38 antibody specifically recognizes the CD4 antigen on most thymocytes, a subpopulation of mature T lymphocytes (ie, MHC class II-restricted T cells, including most T helper cells), monocytes, macrophages, and some dendritic cells. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. The OX-38 antibody has been reported to bind to the same epitope of CD4 as that recognized by W3/25 mAb, which is a different epitope than that recognized by OX-35 mAb. In vivo blocking of some cell-mediated immune responses by mAb OX-38 has been reported. Injection of OX-38 mAb induces allograft unresponsiveness in rats, with varying results depending on the rat strain used (high or low responder). Furthermore, in vivo depletion of CD4+ lymphocytes has been reported with this antibody.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Classification Monoclonal
Clone OX-38
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Cd4, p55, W3/25, T-cell surface antigen T4/Leu-3
Host Species Mouse
Immunogen Rat thymocyte glycoproteins
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.