Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Rabbit anti-Human, Unconjugated, Clone: RM345, Abnova™

Catalog No. 89339201 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-201 Supplier Abnova Corporation Supplier No. MAB21984
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for WB-Ce, IHC-P.

Specifications

Antigen CD4
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM345
Conjugate Unconjugated
Description CD4 molecule
Gene CD4
Gene Alias CD4mut
Host Species Rabbit
Immunogen A peptide corresponding to N-terminus of human CD4
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 920
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.