Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Rat anti-Mouse, FITC, Clone: 4AM15, Abnova™

Catalog No. 89933797 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-797 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-797 Supplier Abnova Corporation Supplier No. MAB12417
Add to Cart
Add to Cart

Rat monoclonal antibody raised against mouse Cd4.

Specifications

Antigen CD4
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 4AM15
Conjugate FITC
Description Rat monoclonal antibody raised against mouse Cd4.
Dilution Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA)
Gene Alias L3T4/Ly-4
Gene Symbols Cd4
Host Species Rat
Immunogen Cd4
Purification Method Protein A/G affinity chromatography
Quantity 50 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12504
Target Species Mouse
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.