Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4 Rat anti-Mouse, FITC, Clone: RM4-5, Abnova™
Rat monoclonal antibody raised against Cd4.
Supplier: Abnova Corporation MAB12209
Specifications
CD4 | |
Monoclonal | |
FITC | |
Flow Cytometry The optimal working dilution should be determined by the end user. | |
L3T4/Ly-4 | |
Rat | |
Affinity Chromatography | |
RUO | |
12504 | |
Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Flow Cytometry | |
RM4-5 | |
Rat monoclonal antibody raised against Cd4. | |
In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer). | |
Cd4 | |
Cd4 | |
100 μg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction