Learn More
Description
Specifications
Specifications
| Antigen | CD4 |
| Applications | Single Cell 3' Sequencing |
| Classification | Monoclonal |
| Clone | Clone GK1.5 |
| Conjugate | Oligo |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3 |
| Host Species | Rat |
| Immunogen | Mouse CTL clone V4 |
| Quantity | 25 Tests |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.