Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4, Rat Anti-Mouse, RB613, GK1.5, BD Horizon™
Mouse Monoclonal Antibody
$138.00 - $346.00
Specifications
Antigen | CD4 |
---|---|
Clone | GK1.5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD4 Monoclonal antibody specifically detects CD4 in Mouse (QC Testing) samples. It is validated for Flow CytometrySpecifications
CD4 | |
0.2 mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3 | |
Mouse CTL clone V4 | |
Primary |
GK1.5 | |
Flow Cytometry | |
RB613 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
12504 | |
IgG2b κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title