Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD4 Recombinant Rabbit Monoclonal Antibody (BLR167J)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $523.50
Specifications
Antigen | CD4 |
---|---|
Clone | BLR167J |
Concentration | 1 mg/mL |
Content And Storage | 4°C |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544519
|
Invitrogen™
MA544519 |
100 μL |
Each for $523.50
|
|
|||||
PIMA544520
|
Invitrogen™
MA544520 |
10 μL |
Each for $173.10
|
|
|||||
Description
Epitope retrieval with Tris-EDTA pH9.0 is recommended for FFPE tissue sections.
The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].Specifications
CD4 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.09% sodium azide; pH 8.2 | |
P06332 | |
12504 | |
Peptide representing a region between residue 407 and the C-terminus (residue 457) of mouse T-cell surface glycoprotein CD4. | |
Antibody |
BLR167J | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Mouse | |
CD4 | |
Activation B7-1 antigen; B7; B7.1; B7-1; BB1; B-lymphocyte activation antigen B7; CD28LG; CD28LG1; CD4; CD4 antigen; CD4 antigen (p55); CD4 antigen p55; Cd4 molecule; CD4 precursor; CD4 receptor; CD4, allele 1; cd4a; CD4mut; CD80; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); CD80 molecule; cell surface glycoprotein CD4; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; fCD4; L3T4; LAB7; Leu-3; Ly-4; lymphocyte antigen CD4; lymphocyte antigen CD4 precursor; membrane protein; p55; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4); T-lymphocyte activation antigen CD80; W3/25; W3/25 antigen | |
CD4 | |
Primary | |
purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title