Learn More
Promega Corporation CD40 Bioassay 5X
Catalog No. PRJA2155 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Type:
Bioassay
Catalog No. Type
PRJA2155 Bioassay
1 options
Catalog No. PRJA2155 Supplier Promega Corporation Supplier No. JA2155
Measure potency and stability of biologics designed to activate CD40 is prequalified according to ICH guidelines. Available in 96- and 384-well plate formats and supplied in a convenient thaw-and-use format, also available in cell propagation model (CPM) format.
  • Measure potency and stability of biologics designed to activate CD40.
  • Prequalified according to ICH guidelines.
  • Available in 96- and 384-well plate formats.
  • Supplied in convenient thaw-and-use format.
  • Also available in cell propagation model (CPM) format.
  • High sensitivity and specificity.
  • Consistent and reproducible results.
  • Suitable for research and clinical applications.
  • Streamlined workflow reduces hands-on time.
  • Reliable and efficient solution for CD40 bioassays.
Specifications
Detection Method Bioluminescence
Type Bioassay
Cell Line CD40 Effector Cells