Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD40 Ligand/TNFSF5 Antibody (hu5c8 (Ruplizumab)) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | CD40 Ligand/TNFSF5 |
---|---|
Clone | hu5c8 (Ruplizumab) |
Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Blocking Assay, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25267825
![]() |
Novus Biologicals
NBP2526780.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP25267802
![]() |
Novus Biologicals
NBP2526780.2MG |
0.2 mg |
Each for $584.50
|
|
|||||
Description
CD40 Ligand/TNFSF5 Monoclonal specifically detects CD40 Ligand/TNFSF5 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
CD40 Ligand/TNFSF5 | |
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5 | |
CD40LG | |
IgG | |
Protein A or G purified | |
The antibody binds to CD40L with an IC50 of 0.845 ug/ml. |
hu5c8 (Ruplizumab) | |
Western Blot, Flow Cytometry, Blocking Assay, CyTOF | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Apoptosis, Asthma, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
959 | |
Human CD40L | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title