Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD40 Ligand/TNFSF5 Antibody (hu5c8 (Ruplizumab)), PerCP, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275907PCP
Description
CD40 Ligand/TNFSF5 Monoclonal specifically detects CD40 Ligand/TNFSF5 in Human samples. It is validated for Flow Cytometry.Specifications
| CD40 Ligand/TNFSF5 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| CD40LG | |
| Human CD40L | |
| 0.1 mL | |
| Primary | |
| The antibody binds to CD40L with an IC50 of 0.845 ug/ml. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| hu5c8 (Ruplizumab) | |
| Flow Cytometry | |
| CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5 | |
| Human | |
| Protein A purified | |
| Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
| 959.0 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction