Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD40 Ligand/TNFSF5 Rabbit anti-Human, Clone: IDEC-131 (Toralizumab), Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | CD40 Ligand/TNFSF5 |
---|---|
Clone | IDEC-131 (Toralizumab) |
Concentration | 1 mg/ml |
Dilution | Immunocytochemistry/Immunofluorescence, Block/Neutralize |
Applications | Immunofluorescence, Neutralization |
Description
CD40 Ligand/TNFSF5 Monoclonal antibody specifically detects CD40 Ligand/TNFSF5 in Human samples. It is validated for Immunofluorescence, NeutralizationSpecifications
CD40 Ligand/TNFSF5 | |
1 mg/ml | |
Immunofluorescence, Neutralization | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
PBS | |
959 | |
IgG κ | |
Protein A purified |
IDEC-131 (Toralizumab) | |
Immunocytochemistry/Immunofluorescence, Block/Neutralize | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5 | |
The original monoclonal antibody was generated by immunization with a soluble fusion protein of human gp39 (gp39-CD8). | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title