Learn More
CD40 Mouse anti-Human, APC, Clone: 5C3, BD
Mouse Monoclonal Antibody
$174.70 - $345.50
Specifications
Antigen | CD40 |
---|---|
Clone | 5C3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561848
|
BD Biosciences
561848 |
25 Tests |
Each for $174.70
|
|
|||||
BDB555591
|
BD Biosciences
555591 |
100 Tests |
Each for $345.50
|
|
|||||
Description
This 5C3 monoclonal antibody specifically binds to CD40, a 45-48 kDa type I integral membrane glycoprotein. CD40 is expressed on B lymphocytes, but is not expressed on terminally differentiated B cells. CD40 is also expressed by endothelial cells, basal epithelial cells and some epithelial cell carcinomas, follicular dendritic cells, macrophages, fibroblasts, keratinocytes, and CD34+ hematopoietic progenitor cells. This antibody is useful for studying the roles played by CD40 in B-cell growth, proliferation, and differentiation including immunoglobulin isotype switching. Anti-CD40 antibodies have been reported to stimulate B-cell proliferation when costimulated with anti-µ, anti-CD20 antibodies or with phorbol esters. 5C3 is capable of inducing B-cell proliferation when presented with IL-4.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD40 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
5C3 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.