Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Mouse anti-Human, APC-R700, Clone: 5C3, BD Horizon™

Catalog No. BDB565179 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565179 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565179 Supplier BD Biosciences Supplier No. 565179
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD40 Monoclonal antibody specifically detects CD40 in Baboon, Cynomolgus, Human, Rhesus Ma samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD40
Applications Flow Cytometry
Classification Monoclonal
Clone 5C3
Conjugate APC-R700
Description Vol. Per test: 5μL
Formulation Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Gene Alias TNFRSF5; TNF receptor superfamily member 5; CD40L receptor; Bp50; p50
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.