Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Rat anti-Human, Clone: FGK45.5, MilliporeSigma™

Catalog No. MABF269025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF269025 25 μL
MABF2690100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF269025 Supplier MilliporeSigma Supplier No. MABF269025UL
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD40 Monoclonal antibody specifically detects CD40 in Human samples. It is validated for Flow Cytometry, Affects Function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD40
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone FGK45.5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG2a in PBS without azide.
Host Species Rat
Immunogen Recombinant mouse CD40 fusion protein.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.