Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD40/TNFRSF5 Antibody (4), Alexa Fluor™ 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289639AF488
Description
CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
CD40/TNFRSF5 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human CD27/ TNFRSF7 Mouse CD27/ TNFRSF7 Human CD30/ TNFRSF8 Human DR6/ TNFRSF21 Human CD137/ 4-1BB Human TNFRSF1B/ CD120b Human HVEM/ TNFRSF14 Mouse HVEM/ TNFRSF14 Human RELT/ TNFRSF19L | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
4 | |
ELISA, Sandwich ELISA Detection | |
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD40/TNFRSF5 extracellular domain (NP_001241.1; Met 1-Arg 193). | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
958 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction