Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD40/TNFRSF5 Antibody (G28.5) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $587.00
Specifications
Antigen | CD40/TNFRSF5 |
---|---|
Clone | G28.5 |
Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25269725
![]() |
Novus Biologicals
NBP2526970.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP25269702
![]() |
Novus Biologicals
NBP2526970.2MG |
0.2 mg |
Each for $587.00
|
|
|||||
Description
CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Functional, CyTOF-ready.Specifications
CD40/TNFRSF5 | |
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
CD40 | |
IgG1 | |
Protein A or G purified | |
48 kD Type I glycoprotein, a member of the TNFR superfamily primarily expressed on normal or malignant B cells or B cell lines, macrophages, follicular dendritic cells, endothelial cells, fibroblasts, and at low levels on plasma cells. |
G28.5 | |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
958 | |
Human erythrocyte-rosette-negative tonsillar lymphocytes. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title