Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD40/TNFRSF5 Antibody (OTI8B8), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270378AF647
Description
CD40/TNFRSF5 Monoclonal specifically detects CD40/TNFRSF5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Product Image.Specifications
CD40/TNFRSF5 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD40 | |
Recombinant protein fragment corresponding to amino acids 21-166 of human CD40/TNFRSF5 (NP_001241) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI8B8 | |
Western Blot, Immunohistochemistry | |
B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
Mouse | |
Affinity Purified | |
Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
958.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction