Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40L (CD154) Mouse anti-Human, BV 510, Clone: 24-31, BD OptiBuild™

Catalog No. BDB752847 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752847 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB752847 Supplier BD Biosciences Supplier No. 752847
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD40L (CD154) Monoclonal antibody specifically detects CD40L (CD154) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD40L (CD154)
Applications Flow Cytometry
Classification Monoclonal
Clone 24-31
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD40 ligand; CD40-L; CD40LG; T-BAM; TNFSF5; TRAP; gp39; hCD40L
Host Species Mouse
Immunogen Human gp39 Protein
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.