Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD41/CD61 Monoclonal Antibody (CO.35E4)
GREENER_CHOICE

Catalog No. PIMA528370
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528370 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528370 Supplier Invitrogen™ Supplier No. MA528370
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse anti Sheep CD41/CD61 antibody, clone CO.35E4 recognizes ruminant CD41/CD61, also known as GpIIb/IIIa. CD41/CD61 is a calcium-complex dependent heterodimer and is expressed almost exclusively on platelets and on their precursors, the megakaryocytes.

CD41 is a transmembrane glycoprotein (integrin family) composed of two chains GPIIb alpha (heavy chain; 120 kDa) and GPIIb beta (light chain; 23 kDa). CD61 non-covalently associates with CD41.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD41/CD61
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone CO.35E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene ITGA2B
Gene Accession No. 0
Gene Alias alphaIIb protein; antigen CD61; BDPLT16; BDPLT2; CD41; CD41B; CD61; glycoprotein GPIIb; glycoprotein GPIIIa; glycoprotein IIb; glycoprotein IIIa; GP2B; GP3A; GPalpha IIb; GPIIb; GPIIIa; GT; GTA; HPA3; INTB3; integrin; integrin alpha 2b; integrin alpha 2b precursor; Integrin alpha-2b; integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; Integrin beta; integrin beta 3; integrin beta 3 subunit; integrin beta3; integrin beta-3; integrin beta-3 subunit; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin subunit beta 3; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41B); integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGA2B; ITGA2B-like protein; ITGAB; ITGB3; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/IIIa complex; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet membrane glycoprotein IIb; Platelet membrane glycoprotein IIIa; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93
Gene Symbols ITGA2B, ITGB3
Host Species Mouse
Immunogen Sheep leucocytes.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008734, 100008735, 100009697, 100009709, 100423047, 101092159, 101100930, 101115719, 102169417, 102183451, 282642, 403788, 403789, 515011
Target Species Canine, Rabbit, Bovine, Sheep, Feline, Horse, Goat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.