Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD41a Monoclonal Antibody (eBioMWReg30 (MWReg30)), NovaFluor™ Blue 555, eBioscience™, Invitrogen™

Catalog No. M044T03B03A
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M044T03B03A 100 μg
M044T02B03A 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M044T03B03A Supplier Invitrogen™ Supplier No. M044T03B03A
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD41, also known as platelet glycoprotein IIb or ITGA2B, is a protein composed of two subunits: a 120 kDa alpha subunit and a 23 kDa beta subunit. It interacts with CD61 (gpIIIa, integrin beta III) in the presence of calcium to form a functional adhesive protein receptor. Initially thought to be expressed exclusively on platelets and megakaryocytes, CD41 is also found on hematopoietic progenitors in the embryo, fetus, and adult, indicating its role in early stages of hematopoietic differentiation. CD41 plays a crucial role in platelet function and blood coagulation by mediating platelet aggregation. Upon blood vessel damage, CD41 binds to various adhesion molecules, including von Willebrand factor, fibrinogen, fibronectin, and vitronectin, facilitating platelet adhesion and aggregation. This receptor is essential for platelet function, contributing to the binding of several adhesion molecules and playing a significant role in hemostasis. Diseases associated with CD41 dysfunction include Glanzmann Thrombasthenia and Platelet type-16 bleeding disorder, highlighting its importance in normal platelet function and blood coagulation processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD41a
Applications Flow Cytometry
Classification Monoclonal
Clone eBioMWReg30 (MWReg30)
Concentration 0.1 mg/mL
Conjugate NovaFluor Blue 555
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Itga2b
Gene Accession No. Q9QUM0
Gene Alias AI172977; alpha IIb; alphaIIb; alphaIIb protein; BDPLT16; BDPLT2; CD41; CD41B; form 1; form 2; GP IIb; GP2B; GPalpha IIb; GPIIb; GT; GTA; HPA3; integrin alpha 2b; integrin alpha 2b (Cd41b); integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin, alpha 2B; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); ITGA2B; ITGAB; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/II Ia complex; platelet glycoprotein IIb of IIb/IIIa complex; platelet membrane glycoprotein IIb; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93
Gene Symbols Itga2b
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16399
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.