Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD41a Monoclonal Antibody (HIP8), NovaFluor™ Blue 585, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD41a |
---|---|
Clone | HIP8 |
Concentration | 0.6 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H059T03B04A
![]() |
Invitrogen™
H059T03B04A |
100 Tests |
Each for $430.00
|
|
|||||
H059T02B04A
![]() |
Invitrogen™
H059T02B04A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD41a Monoclonal antibody specifically detects CD41a in Human samples. It is validated for Flow CytometrySpecifications
CD41a | |
0.6 μg/Test | |
Monoclonal | |
Mouse | |
Human | |
P08514 | |
3674 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
HIP8 | |
Flow Cytometry | |
NovaFluor Blue 585 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI172977; alpha IIb; alphaIIb; alphaIIb protein; BDPLT16; BDPLT2; CD41; CD41B; form 1; form 2; GP IIb; GP2B; GPalpha IIb; GPIIb; GT; GTA; HPA3; integrin alpha 2b; integrin alpha 2b (Cd41b); integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin, alpha 2B; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); ITGA2B; ITGAB; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/II Ia complex; platelet glycoprotein IIb of IIb/IIIa complex; platelet membrane glycoprotein IIb; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93 | |
IgG1 κ | |
Affinity Chromatography | |
Itga2b |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title