Learn More
Description
The HIP8 monoclonal antibody specifically binds to the α-chain of CD41. CD41 is also known as Integrin αIIb or Platelet GPIIb. The calcium-dependent complex of CD41 and CD61 (β3 integrin or GPIIIa) is normally expressed on platelets and megakaryocytes. The CD41/CD61 complex is the receptor for fibrinogen, fibronectin and von Willebrand factor, and mediates platelet adhesion and aggregation. CD41 (clone HIP8) completely inhibits ADP-, epinephrine-, and collagen-induced platelet activation, and partially inhibits ristocetin- and thrombin-induced platelet activation. This antibody is useful in the morphological and physiological studies of platelets and megakaryocytes.
Specifications
Specifications
| Antigen | CD41a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HIP8 |
| Conjugate | APC |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Purified platelet membrane glycoproteins |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.