Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD41a Mouse anti-Human, APC, Clone: HIP8, BD

Catalog No. BDB561852 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561852 25 Tests
BDB559777 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561852 Supplier BD Biosciences Supplier No. 561852
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HIP8 monoclonal antibody specifically binds to the α-chain of CD41. CD41 is also known as Integrin αIIb or Platelet GPIIb. The calcium-dependent complex of CD41 and CD61 (β3 integrin or GPIIIa) is normally expressed on platelets and megakaryocytes. The CD41/CD61 complex is the receptor for fibrinogen, fibronectin and von Willebrand factor, and mediates platelet adhesion and aggregation. CD41 (clone HIP8) completely inhibits ADP-, epinephrine-, and collagen-induced platelet activation, and partially inhibits ristocetin- and thrombin-induced platelet activation. This antibody is useful in the morphological and physiological studies of platelets and megakaryocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD41a
Applications Flow Cytometry
Classification Monoclonal
Clone HIP8
Conjugate APC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified platelet membrane glycoproteins
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.