Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD42a Monoclonal Antibody (1E2D10)
GREENER_CHOICE

Catalog No. FERMA548470
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548470 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548470 Supplier Invitrogen™ Supplier No. MA548470
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD42a Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

Single-chain membrane glycoprotein that forms a non-covalent complex with GPIb. (MW 23 kDa) Reactivity with resting and activated platelets, weakly on monocytes, megakaryocytes and attachment site for the platelet plasma membrane to the submembrane cytoskeleton. GPIb/IX complex, functions as the receptor for ristocetin-induced binding of von Willebrand factor and as the von Willebrand factor-depend adhesion receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD42a
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1E2D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GP9
Gene Accession No. O88186, P14770
Gene Alias CD42a; Glycoprotein 9; glycoprotein 9 (platelet); glycoprotein IX (platelet); glycoprotein IX platelet; GP9; GPIX; GP-IX; platelet glycoprotein IX; Platelet GPIX
Gene Symbols GP9
Host Species Mouse
Immunogen Purified recombinant fragment of human CD42A (AA: extra(17-147)) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2815, 54368
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.