Learn More
Invitrogen™ CD42a Monoclonal Antibody (GR-P)

Description
This antibody recognizes an extracellular epitope of CD42a (glycoprotein 9), a 22 kDa transmembrane protein constitutively expressed on megakaryocytes and platelets.
Specifications
Specifications
| Antigen | CD42a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | GR-P |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | GP9 |
| Gene Accession No. | P14770 |
| Gene Alias | CD42a; Glycoprotein 9; glycoprotein 9 (platelet); glycoprotein IX (platelet); glycoprotein IX platelet; GP9; GPIX; GP-IX; platelet glycoprotein IX; Platelet GPIX |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.