Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD42a Mouse anti-Human, PE, Clone: ALMA.16, BD

Catalog No. BDB561853 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561853 25 Tests
BDB558819 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561853 Supplier BD Biosciences Supplier No. 561853
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 17-22kD, single chain integral membrane glycoprotein, also known as GPIX. CD42a forms a noncovalently linked complex (GPIb/GPIX/GPV) with CD42b, CD42c and CD42d. It is expressed on platelets and megakaryocytes and is absent on the platelets of patients with Bernadr-Soulier Syndrome (BSS). Although the CD42a function is not fully understood, GPIX glycoprotein is important for the assembly and membrane expression of the complex and for the maintenance of the functional conformation of CD42b (GPIb).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD42a
Applications Flow Cytometry
Classification Monoclonal
Clone ALMA.16
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.