Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD42a Mouse, anti-Human, R718, Clone: ALMA.16, BD OptiBuild™

Catalog No. BDB752068 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752068 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752068 Supplier BD Biosciences Supplier No. 752068
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ALMA.16 monoclonal antibody specifically recognizes CD42a. CD42a is a 17-22 kDa type I transmembrane glycoprotein that is also known as Platelet glycoprotein IX (GPIX), or Glycoprotein 9 (GP9). CD42a forms a noncovalently linked complex (GPIb/GPIX/GPV) with CD42b, CD42c and CD42d that may serve as a receptor for von Willebrand factor. It is expressed on platelets and megakaryocytes and is absent on the platelets of patients with Bernadr-Soulier Syndrome (BSS). Although the CD42a function is not fully understood, GPIX glycoprotein is important for the assembly and membrane expression of the CD42 complex and for the maintenance of the functional conformation of CD42b (GPIb).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD42a
Applications Flow Cytometry
Classification Monoclonal
Clone ALMA.16
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.