Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD42a Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584587
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584587 Supplier Invitrogen™ Supplier No. PA584587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HGLTALPALP ARTRHLLLAN NSLQSVPPGA FDHLPQLQTL DVTQNPWHCD C Highest antigen sequence indentity to the following orthologs: Rat - 76%, Mouse - 82%.

Single-chain membrane glycoprotein that forms a non-covalent complex with GPIb. (MW 23 kDa) Reactivity with resting and activated platelets, weakly on monocytes, megakaryocytes and attachment site for the platelet plasma membrane to the submembrane cytoskeleton. GPIb/IX complex, functions as the receptor for ristocetin-induced binding of von Willebrand factor and as the von Willebrand factor-depend adhesion receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD42a
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GP9
Gene Accession No. P14770
Gene Alias CD42a; Glycoprotein 9; glycoprotein 9 (platelet); glycoprotein IX (platelet); glycoprotein IX platelet; GP9; GPIX; GP-IX; platelet glycoprotein IX; Platelet GPIX
Gene Symbols GP9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GP9. Recombinant protein control fragment (Product #RP-103494).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2815
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.