Learn More
Invitrogen™ CD42a Polyclonal Antibody

Description
Positive test controls include: K-562. The target is usually found in the following locations: Membrane, Single-pass type I membrane protein. Immunogen sequence: TKDCPSPCTC RALETMGLWV DCRGHGLTAL PALPARTRHL LLANNSLQSV PPGAFDHLPQ LQTLDVTQNP WHCDCSLTYL RLWLEDRTPE ALLQVRCASP SLAAHGPLGR LTGYQLGSCG WQLQASWVRP G.
Specifications
Specifications
| Antigen | CD42a |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.31 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GP9 |
| Gene Accession No. | O88186, P14770 |
| Gene Alias | CD42a; Glycoprotein 9; glycoprotein 9 (platelet); glycoprotein IX (platelet); glycoprotein IX platelet; GP9; GPIX; GP-IX; platelet glycoprotein IX; Platelet GPIX |
| Gene Symbols | GP9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.