Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD42b (GPIb) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD42b (GPIb) Ab-1, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS1174P1

 View more versions of this product

Catalog No. MS1174P1


Only null left
Add to Cart

Description

Description

The CD42b glycoprotein, also known as GPIb, is a cofactor of ristocetin-induced aggregation and is involved in the binding of platelets to blood vessel walls. The CD42b antigen is expressed on platelets and on megakaryocytes in bone marrow. The absence of CD42b antigen on platelets may indicate Bernard-Soulier disease.

Host Species: Mouse

Clone: 42C01

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Immunogen: Recombinant CD42b.

Molecular Weight: 135-145 kDa (α-chain) and 22-25 kDa (β-chain)

Positive Control: Tonsil

Cellular Localization: Cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD42b Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Hematopoietic
Human
IgG1
Immunohistochemistry (Paraffin)
42C01
Unconjugated
Recombinant CD42b
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.