Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD42b/GPIb alpha Antibody (AK2), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063790APCCY7
Description
CD42b/GPIb alpha Monoclonal specifically detects CD42b/GPIb alpha in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
CD42b/GPIb alpha | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
GP1BA | |
Made to Human CD42b | |
0.1 mL | |
Primary | |
Recognises the human CD42b cell surface antigen, also known as platelet glycoprotein GP1B. CD42b is expressed by platelets and megakaryocytes. Clone AK2 has been reported to block the binding of von Willebrand Factor (VWF) to platelets (1). | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
AK2 | |
Flow Cytometry | |
Antigen CD42b-alpha, BSS, CD42b, CD42b antigen, CD42b-alpha, glycoprotein Ib (platelet), alpha polypeptide, Glycoprotein Ibalpha, GP1B, GP-Ib alpha, GPIbA, GPIb-alpha, MGC34595, platelet glycoprotein Ib alpha chain, platelet membrane glycoprotein 1b-alpha subunit | |
Mouse | |
Protein G purified | |
Immunology | |
2811.0 | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction